Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACCACCTAAAGAGGCACATCCTTA[C/T]TCACACTGGTAAGTCTCTTGTTTAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616569 MIM: 147559 | ||||||||||||||||||||
Literature Links: |
CSAD PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CSAD - cysteine sulfinic acid decarboxylase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ITGB7 - integrin subunit beta 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF740 - zinc finger protein 740 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001004304.3 | 959 | Missense Mutation | ACT,ATT | T,I 122 | NP_001004304.1 | |
XM_006719344.2 | 959 | Missense Mutation | ACT,ATT | T,I 122 | XP_006719407.1 | |
XM_011538194.2 | 959 | Missense Mutation | ACT,ATT | T,I 122 | XP_011536496.1 | |
XM_017019181.1 | 959 | Missense Mutation | ACT,ATT | T,I 122 | XP_016874670.1 |