Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCATTGATAAGGCCAAGGTCTTGGG[C/T]TTCTGCCAGGATCATGAGGGCACAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608730 | ||||||||||||||||||||
Literature Links: |
ANKRD52 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ANKRD52 - ankyrin repeat domain 52 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173595.3 | 2932 | Missense Mutation | ACC,GCC | T,A 948 | NP_775866.2 | |
XM_011538197.2 | 2932 | Missense Mutation | ACC,GCC | T,A 887 | XP_011536499.1 | |
XM_017019183.1 | 2932 | Missense Mutation | ACC,GCC | T,A 947 | XP_016874672.1 | |
XM_017019184.1 | 2932 | Missense Mutation | ACC,GCC | T,A 948 | XP_016874673.1 |
SLC39A5 - solute carrier family 39 member 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |