Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACTCACCTGAAGCAGGATGTAAGC[A/G]TATAATTTGTCAGACGGGCAATATG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
C12orf77 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C12orf77 - chromosome 12 open reading frame 77 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001101339.2 | 344 | Missense Mutation | ACG,ATG | T,M 43 | NP_001094809.2 | |
XM_017018966.1 | 344 | Missense Mutation | ACG,ATG | T,M 115 | XP_016874455.1 |
LOC645177 - uncharacterized LOC645177 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |