Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTGTCAAAGAGTCCATCTGTTGCC[A/G]TCAGGATAATGTCTCCTAGCTGGAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609668 MIM: 608368 | ||||||||||||||||||||
Literature Links: |
PPTC7 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PPTC7 - PTC7 protein phosphatase homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_139283.1 | 887 | Missense Mutation | ACG,ATG | T,M 220 | NP_644812.1 |
RAD9B - RAD9 checkpoint clamp component B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |