Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCAGCGCCCTGGCCATAACCGTAG[A/C]CTTTTGGCCCATACTTCTTTCCGTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601871 MIM: 607799 | ||||||||||||||||||||
Literature Links: |
CSRP2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CSRP2 - cysteine and glycine rich protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300965.1 | 313 | Intron | NP_001287894.1 | |||
NM_001321.2 | 313 | Missense Mutation | GGC,GTC | G,V 70 | NP_001312.1 |
ZDHHC17 - zinc finger DHHC-type containing 17 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |