Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCCTTCTAGCTGCCCACTATTTCC[C/T]ACTGCCTTGGGTTTATAGACATCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605378 MIM: 606633 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AAAS PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AAAS - aladin WD repeat nucleoporin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SP7 - Sp7 transcription factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001173467.2 | 1025 | Silent Mutation | GTA,GTG | V,V 234 | NP_001166938.1 | |
NM_001300837.1 | 1025 | Silent Mutation | GTA,GTG | V,V 216 | NP_001287766.1 | |
NM_152860.1 | 1025 | Silent Mutation | GTA,GTG | V,V 234 | NP_690599.1 | |
XM_011537900.2 | 1025 | Silent Mutation | GTA,GTG | V,V 216 | XP_011536202.1 |