Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCACACGACAGATTTCATGGTTGT[C/T]CTTCCTCCAAACCGTAAGTTGACTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611549 | ||||||||||||||||||||
Literature Links: |
NALCN PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NALCN - sodium leak channel, non-selective | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_052867.2 | 4409 | Silent Mutation | AGA,AGG | R,R 1693 | NP_443099.1 | |
XM_011521067.2 | 4409 | Silent Mutation | AGA,AGG | R,R 1712 | XP_011519369.1 | |
XM_011521068.2 | 4409 | Silent Mutation | AGA,AGG | R,R 1693 | XP_011519370.1 | |
XM_011521069.2 | 4409 | Silent Mutation | AGA,AGG | R,R 1683 | XP_011519371.1 | |
XM_017020534.1 | 4409 | Silent Mutation | AGA,AGG | R,R 1664 | XP_016876023.1 | |
XM_017020535.1 | 4409 | Silent Mutation | AGA,AGG | R,R 1664 | XP_016876024.1 | |
XM_017020536.1 | 4409 | Silent Mutation | AGA,AGG | R,R 1544 | XP_016876025.1 | |
XM_017020537.1 | 4409 | Silent Mutation | AGA,AGG | R,R 1438 | XP_016876026.1 |
NALCN-AS1 - NALCN antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |