Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTGATGATGACATTGAAGTTTTTC[A/G]ATTACAAGCCCCAGTATGGGAATTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608102 MIM: 605653 | ||||||||||||||||||||
Literature Links: |
CLN5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CLN5 - ceroid-lipofuscinosis, neuronal 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006493.2 | 462 | Missense Mutation | CAA,CGA | Q,R 145 | NP_006484.1 | |
XM_011534917.2 | 462 | Missense Mutation | CAA,CGA | Q,R 145 | XP_011533219.1 |
FBXL3 - F-box and leucine rich repeat protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012158.2 | 462 | Intron | NP_036290.1 | |||
XM_005266336.1 | 462 | Intron | XP_005266393.1 | |||
XM_005266337.2 | 462 | Intron | XP_005266394.1 | |||
XM_017020538.1 | 462 | Intron | XP_016876027.1 |