Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACAGAAGAATGGAAGCATTCCAGG[A/G]ACTTCGTAAACCATCAGCACGTTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616327 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHAMP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CHAMP1 - chromosome alignment maintaining phosphoprotein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164144.1 | 349 | Missense Mutation | GAA,GGA | E,G 6 | NP_001157616.1 | |
NM_001164145.1 | 349 | Missense Mutation | GAA,GGA | E,G 6 | NP_001157617.1 | |
NM_032436.2 | 349 | Missense Mutation | GAA,GGA | E,G 6 | NP_115812.1 |
LINC01054 - long intergenic non-protein coding RNA 1054 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |