Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAAATGTGAGTTTTATATGGATCA[C/G]CAATAGCATTCTCTGAGGCGGAGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616066 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC169-SOHLH2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCDC169-SOHLH2 - CCDC169-SOHLH2 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198910.1 | 1580 | Missense Mutation | GCT,GGT | A,G 422 | NP_001185839.1 |
SOHLH2 - spermatogenesis and oogenesis specific basic helix-loop-helix 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282147.1 | 1580 | Intron | NP_001269076.1 | |||
NM_017826.2 | 1580 | Missense Mutation | GCT,GGT | A,G 345 | NP_060296.2 |