Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTCTTCTGTTACCAATTGTATAGG[A/G]GATGTTAGAAGCTCTCAAGGCACTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606977 | ||||||||||||||||||||
Literature Links: |
COG6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
COG6 - component of oligomeric golgi complex 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145079.1 | 255 | Missense Mutation | GAG,GGG | E,G 52 | NP_001138551.1 | |
NM_020751.2 | 255 | Missense Mutation | GAG,GGG | E,G 52 | NP_065802.1 | |
XM_011535168.1 | 255 | Missense Mutation | GAG,GGG | E,G 52 | XP_011533470.1 |
MIR4305 - microRNA 4305 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |