Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGGGTAAGAAATACAAATGAGACC[C/T]CGAACAAAATCATCACCATCATCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC168 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCDC168 - coiled-coil domain containing 168 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TEX30 - testis expressed 30 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286775.1 | 629 | Intron | NP_001273704.1 | |||
NM_001286776.1 | 629 | Silent Mutation | CGA,CGG | R,R 84 | NP_001273705.1 | |
NM_138779.4 | 629 | Silent Mutation | CGA,CGG | R,R 125 | NP_620134.3 | |
XM_005254097.2 | 629 | Silent Mutation | CGA,CGG | R,R 125 | XP_005254154.1 | |
XM_011521135.2 | 629 | Silent Mutation | CGA,CGG | R,R 125 | XP_011519437.1 | |
XM_011521136.2 | 629 | Silent Mutation | CGA,CGG | R,R 81 | XP_011519438.1 | |
XM_017020852.1 | 629 | Silent Mutation | CGA,CGG | R,R 125 | XP_016876341.1 |