Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTCTTTGTTCAAAGTGTCTCTTTC[C/T]TGGGTCTCTTGAGTTTCTTTATATG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600860 | ||||||||||||||||||||
Literature Links: |
GTF3A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GTF3A - general transcription factor IIIA | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002097.2 | 1662 | Intron | NP_002088.2 |
MTIF3 - mitochondrial translational initiation factor 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166261.1 | 1662 | Intron | NP_001159733.1 | |||
NM_001166262.1 | 1662 | Intron | NP_001159734.1 | |||
NM_001166263.1 | 1662 | Silent Mutation | CAA,CAG | Q,Q 258 | NP_001159735.1 | |
NM_152912.4 | 1662 | Intron | NP_690876.3 | |||
XM_006719771.3 | 1662 | Silent Mutation | CAA,CAG | Q,Q 258 | XP_006719834.1 | |
XM_006719772.3 | 1662 | Silent Mutation | CAA,CAG | Q,Q 258 | XP_006719835.1 | |
XM_011534957.2 | 1662 | Silent Mutation | CAA,CAG | Q,Q 258 | XP_011533259.1 | |
XM_011534958.2 | 1662 | Silent Mutation | CAA,CAG | Q,Q 258 | XP_011533260.1 | |
XM_011534959.2 | 1662 | Silent Mutation | CAA,CAG | Q,Q 258 | XP_011533261.1 | |
XM_011534960.2 | 1662 | Silent Mutation | CAA,CAG | Q,Q 258 | XP_011533262.1 | |
XM_011534961.2 | 1662 | Silent Mutation | CAA,CAG | Q,Q 258 | XP_011533263.1 | |
XM_011534962.2 | 1662 | Silent Mutation | CAA,CAG | Q,Q 258 | XP_011533264.1 | |
XM_011534963.2 | 1662 | Silent Mutation | CAA,CAG | Q,Q 258 | XP_011533265.1 | |
XM_017020417.1 | 1662 | Intron | XP_016875906.1 |