Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGCAGGTAACGCGTTGGGTTCTTT[C/G]CATTGCCGACCGCATTTTACTATCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
CLDN10 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CLDN10 - claudin 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001160100.1 | 412 | Intron | NP_001153572.1 | |||
NM_006984.4 | 412 | Intron | NP_008915.1 | |||
NM_182848.3 | 412 | Missense Mutation | TTC,TTG | F,L 59 | NP_878268.1 | |
XM_011521134.2 | 412 | Intron | XP_011519436.1 | |||
XM_017020843.1 | 412 | Missense Mutation | TTC,TTG | F,L 59 | XP_016876332.1 | |
XM_017020844.1 | 412 | Missense Mutation | TTC,TTG | F,L 59 | XP_016876333.1 |