Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAATTCTTCTCGGTGGCTACCGTG[C/T]TCAAGGTCTGCAGGTAGCGCAGCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608327 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
STOML3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
STOML3 - stomatin like 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001144033.1 | 983 | Missense Mutation | AAC,AGC | N,S 245 | NP_001137505.1 | |
NM_145286.2 | 983 | Missense Mutation | AAC,AGC | N,S 254 | NP_660329.1 | |
XM_017020400.1 | 983 | Missense Mutation | AAC,AGC | N,S 183 | XP_016875889.1 |