Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGGATGGCTCTTGCCCTGGGACAC[A/G]GCGATGTCCACGGCCGCCAGCGCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601632 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC107987188 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC107987188 - uncharacterized LOC107987188 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
POU4F1 - POU class 4 homeobox 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006237.3 | 435 | Silent Mutation | GCC,GCT | A,A 67 | NP_006228.3 |
RNF219-AS1 - RNF219 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |