Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCTGAACCGAAACTAGCTACCTGA[C/T]GATATGAGGCAAAAGACAAGGAAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607519 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PARP4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PARP4 - poly(ADP-ribose) polymerase family member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006437.3 | 4357 | Missense Mutation | CAT,CGT | H,R 1345 | NP_006428.2 | |
XM_011534931.1 | 4357 | Missense Mutation | CAT,CGT | H,R 1392 | XP_011533233.1 | |
XM_011534932.2 | 4357 | Missense Mutation | CAT,CGT | H,R 1345 | XP_011533234.1 |