Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGACCGGGGCTTTCCATGGGCTAC[A/G]GGGTTTGAGGAGATTGCATCTAAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609680 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR4500HG PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MIR4500HG - MIR4500 host gene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLITRK5 - SLIT and NTRK like family member 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015567.1 | 1276 | Missense Mutation | CAG,CGG | Q,R 129 | NP_056382.1 | |
XM_005254038.4 | 1276 | Missense Mutation | CAG,CGG | Q,R 129 | XP_005254095.1 | |
XM_005254039.2 | 1276 | Missense Mutation | CAG,CGG | Q,R 129 | XP_005254096.1 |