Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGTCCACATCTGTGAACGTTTTCA[G/T]GCTAAATCTGGCCATTTCAGATCTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605666 | ||||||||||||||||||||
Literature Links: |
CYSLTR2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CYSLTR2 - cysteinyl leukotriene receptor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308465.1 | 931 | Missense Mutation | AGG,ATG | R,M 77 | NP_001295394.1 | |
NM_001308467.1 | 931 | Missense Mutation | AGG,ATG | R,M 77 | NP_001295396.1 | |
NM_001308468.1 | 931 | Missense Mutation | AGG,ATG | R,M 77 | NP_001295397.1 | |
NM_001308469.1 | 931 | Missense Mutation | AGG,ATG | R,M 77 | NP_001295398.1 | |
NM_001308470.1 | 931 | Missense Mutation | AGG,ATG | R,M 77 | NP_001295399.1 | |
NM_001308471.1 | 931 | Missense Mutation | AGG,ATG | R,M 77 | NP_001295400.1 | |
NM_001308476.1 | 931 | Missense Mutation | AGG,ATG | R,M 77 | NP_001295405.1 | |
NM_020377.3 | 931 | Missense Mutation | AGG,ATG | R,M 77 | NP_065110.1 |