Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGTATCCAATTTTGTTTATAGTCC[A/T]GTTACAACACAGGGATCACAACAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605553 | ||||||||||||||||||||
Literature Links: |
PAPOLA PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PAPOLA - poly(A) polymerase alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001252006.1 | 229 | Silent Mutation | NP_001238935.1 | |||
NM_001252007.1 | 229 | Silent Mutation | NP_001238936.1 | |||
NM_001293627.1 | 229 | Silent Mutation | NP_001280556.1 | |||
NM_001293628.1 | 229 | UTR 5 | NP_001280557.1 | |||
NM_001293632.1 | 229 | UTR 5 | NP_001280561.1 | |||
NM_032632.4 | 229 | Silent Mutation | NP_116021.2 | |||
XM_005267281.1 | 229 | Silent Mutation | XP_005267338.1 | |||
XM_005267282.2 | 229 | Silent Mutation | XP_005267339.1 | |||
XM_017020935.1 | 229 | Silent Mutation | XP_016876424.1 | |||
XM_017020936.1 | 229 | Silent Mutation | XP_016876425.1 |