Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCCTTTCTAGGAGCTGCTGCCGC[C/T]GGGCGTCCCCCGTGGCCTCGGGCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612800 MIM: 615910 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CARS2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CARS2 - cysteinyl-tRNA synthetase 2, mitochondrial (putative) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024537.3 | 1350 | Missense Mutation | NP_078813.1 | |||
XM_006719953.2 | 1350 | Missense Mutation | XP_006720016.1 | |||
XM_011521116.1 | 1350 | Missense Mutation | XP_011519418.1 | |||
XM_011521117.2 | 1350 | Intron | XP_011519419.1 | |||
XM_011521118.2 | 1350 | Intron | XP_011519420.1 | |||
XM_017020741.1 | 1350 | Missense Mutation | XP_016876230.1 | |||
XM_017020742.1 | 1350 | Intron | XP_016876231.1 |
NAXD - NAD(P)HX dehydratase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |