Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTCCTCCAAGGACTGTTTTTCTT[C/T]AAATGACTGGCACCAGCAGCATAAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 131244 | ||||||||||||||||||||
Literature Links: |
EDNRB PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
EDNRB - endothelin receptor type B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000115.3 | 1648 | Missense Mutation | AAA,GAA | K,E 409 | NP_000106.1 | |
NM_001122659.2 | 1648 | Missense Mutation | AAA,GAA | K,E 409 | NP_001116131.1 | |
NM_001201397.1 | 1648 | Missense Mutation | AAA,GAA | K,E 499 | NP_001188326.1 | |
NM_003991.3 | 1648 | Intron | NP_003982.1 |
EDNRB-AS1 - EDNRB antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |