Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCAGCCGGGCCCTGCAGCTCTCGC[A/G]GAGCTTGGCGACGGTGGCCATGGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 153330 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GRTP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GRTP1 - growth hormone regulated TBC protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286732.1 | 2291 | Intron | NP_001273661.1 | |||
NM_001286733.1 | 2291 | Missense Mutation | CCG,CTG | P,L 205 | NP_001273662.1 | |
NM_024719.3 | 2291 | Missense Mutation | CGC,TGC | R,C 325 | NP_078995.2 | |
XM_005268333.3 | 2291 | UTR 3 | XP_005268390.1 | |||
XM_011537521.2 | 2291 | Intron | XP_011535823.1 | |||
XM_011537522.2 | 2291 | Intron | XP_011535824.1 | |||
XM_017020755.1 | 2291 | UTR 3 | XP_016876244.1 | |||
XM_017020756.1 | 2291 | UTR 3 | XP_016876245.1 |
LAMP1 - lysosomal associated membrane protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |