Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTTCCATCAGGTCCGTATGAAAAC[A/G]TGGGTGCGCGGAATGATTCTGTTAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIPEPP3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MIPEPP3 - mitochondrial intermediate peptidase pseudogene 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZDHHC20 - zinc finger DHHC-type containing 20 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286638.1 | 363 | Intron | NP_001273567.1 | |||
NM_153251.3 | 363 | Intron | NP_694983.2 | |||
XM_006719795.1 | 363 | Missense Mutation | ACG,ATG | T,M 249 | XP_006719858.1 | |
XM_006719796.1 | 363 | Missense Mutation | ACG,ATG | T,M 249 | XP_006719859.1 | |
XM_006719797.1 | 363 | Missense Mutation | ACG,ATG | T,M 249 | XP_006719860.1 | |
XM_011535030.2 | 363 | Missense Mutation | ACG,ATG | T,M 101 | XP_011533332.1 | |
XM_017020525.1 | 363 | Intron | XP_016876014.1 | |||
XM_017020526.1 | 363 | Missense Mutation | ACG,ATG | T,M 170 | XP_016876015.1 | |
XM_017020527.1 | 363 | Missense Mutation | ACG,ATG | T,M 170 | XP_016876016.1 |