Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAACACCGAGATGGACACCCTGCT[C/G]GTGTCTAGGTAGGAGTTGGAGTGCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609678 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLITRK1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLITRK1 - SLIT and NTRK like family member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001281503.1 | 2722 | Silent Mutation | ACC,ACG | T,T 612 | NP_001268432.1 | |
NM_052910.2 | 2722 | Silent Mutation | ACC,ACG | T,T 612 | NP_443142.1 |