Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTAGGTAAGGCATGAGCACCCTAGG[C/T]TTCTTCTGTGTATCTCTTGCTGCTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 191275 | ||||||||||||||||||||
Literature Links: |
DCT PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DCT - dopachrome tautomerase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001129889.2 | 1982 | Missense Mutation | ACC,GCC | T,A 552 | NP_001123361.1 | |
NM_001322182.1 | 1982 | Missense Mutation | ACC,GCC | T,A 208 | NP_001309111.1 | |
NM_001322183.1 | 1982 | Missense Mutation | ACC,GCC | T,A 208 | NP_001309112.1 | |
NM_001322184.1 | 1982 | Missense Mutation | ACC,GCC | T,A 208 | NP_001309113.1 | |
NM_001322185.1 | 1982 | Missense Mutation | ACC,GCC | T,A 208 | NP_001309114.1 | |
NM_001322186.1 | 1982 | Missense Mutation | ACC,GCC | T,A 456 | NP_001309115.1 | |
NM_001922.4 | 1982 | Missense Mutation | ACC,GCC | T,A 519 | NP_001913.2 | |
XM_011521049.1 | 1982 | Missense Mutation | ACC,GCC | T,A 456 | XP_011519351.1 | |
XM_017020401.1 | 1982 | Missense Mutation | ACC,GCC | T,A 562 | XP_016875890.1 |