Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCCAGGAGGTGGTGAATATGGCCG[T/G]GATCGTGCCCAAAGAGGAGGGCGTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610418 | ||||||||||||||||||||
Literature Links: |
WDFY2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
WDFY2 - WD repeat and FYVE domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_052950.3 | 323 | Missense Mutation | GGG,GTG | G,V 31 | NP_443182.1 | |
XM_011534914.1 | 323 | Missense Mutation | GGG,GTG | G,V 31 | XP_011533216.1 | |
XM_011534915.2 | 323 | UTR 5 | XP_011533217.1 | |||
XM_017020386.1 | 323 | UTR 5 | XP_016875875.1 |