Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCTATCTTTTTGTGTGAGGATCTG[A/C]AGGTAAAGTATTAATCTTATATAGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 163905 | ||||||||||||||||||||
Literature Links: |
HMGB1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HMGB1 - high mobility group box 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
USPL1 - ubiquitin specific peptidase like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321532.1 | 936 | UTR 5 | NP_001308461.1 | |||
NM_001321533.1 | 936 | UTR 5 | NP_001308462.1 | |||
NM_001321534.1 | 936 | Intron | NP_001308463.1 | |||
NM_005800.4 | 936 | Missense Mutation | AAG,CAG | K,Q 76 | NP_005791.3 | |
XM_005266213.3 | 936 | Missense Mutation | AAG,CAG | K,Q 76 | XP_005266270.1 | |
XM_006719751.3 | 936 | Missense Mutation | AAG,CAG | K,Q 70 | XP_006719814.1 | |
XM_017020314.1 | 936 | Missense Mutation | AAG,CAG | K,Q 76 | XP_016875803.1 | |
XM_017020315.1 | 936 | UTR 5 | XP_016875804.1 | |||
XM_017020316.1 | 936 | UTR 5 | XP_016875805.1 |