Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTCCCTGGAGGTCATCTGGAGTTC[C/G]GGTGAGCAGCCGCGGACGCGAGGAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615792 MIM: 603921 | ||||||||||||||||||||
Literature Links: |
NUDT15 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NUDT15 - nudix hydrolase 15 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304745.1 | 70 | Missense Mutation | CGT,GGT | R,G 53 | NP_001291674.1 | |
NM_018283.3 | 70 | Missense Mutation | CGT,GGT | R,G 53 | NP_060753.1 |
SUCLA2 - succinate-CoA ligase ADP-forming beta subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003850.2 | 70 | Intron | NP_003841.1 | |||
XM_017020806.1 | 70 | UTR 5 | XP_016876295.1 | |||
XM_017020807.1 | 70 | Intron | XP_016876296.1 | |||
XM_017020808.1 | 70 | Intron | XP_016876297.1 | |||
XM_017020809.1 | 70 | Intron | XP_016876298.1 |