Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGGCAGGCTGCGTCCCACTGCTCC[A/T]GGGCCTGGTCCTGGTCCTCGCCCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613872 MIM: 613713 MIM: 176895 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
F10 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
F10 - coagulation factor X | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC102724474 - WAS/WASL-interacting protein family member 2-like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PCID2 - PCI domain containing 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PROZ - protein Z, vitamin K dependent plasma glycoprotein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256134.1 | 33 | Missense Mutation | CAG,CTG | Q,L 9 | NP_001243063.1 | |
NM_003891.2 | 33 | Missense Mutation | CAG,CTG | Q,L 9 | NP_003882.1 | |
XM_017020812.1 | 33 | Missense Mutation | CAG,CTG | Q,L 54 | XP_016876301.1 | |
XM_017020813.1 | 33 | Missense Mutation | CAG,CTG | Q,L 9 | XP_016876302.1 |