Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGTTTTCCTGGGAGCAGGGTTCAT[A/G]CATATGACTGCTGAAGCCCTGGAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616091 MIM: 612166 | ||||||||||||||||||||
Literature Links: |
LOC101929718 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC101929718 - uncharacterized LOC101929718 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
METTL17 - methyltransferase like 17 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC39A2 - solute carrier family 39 member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256588.1 | 378 | Missense Mutation | ATA,ATG | I,M 62 | NP_001243517.1 | |
NM_014579.3 | 378 | Missense Mutation | ATA,ATG | I,M 62 | NP_055394.2 |