Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTGCATCCCAAGGGGCAGACTCAC[C/T]TGTGGTAGTCGGTGACACAGTATAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609066 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AJUBA PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AJUBA - ajuba LIM protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001289097.1 | 1746 | Intron | NP_001276026.1 | |||
NM_032876.5 | 1746 | Missense Mutation | AAA,AGA | K,R 457 | NP_116265.1 | |
NM_198086.2 | 1746 | Missense Mutation | AAA,AGA | K,R 40 | NP_932352.1 |