Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGAGCTGCTGCTCTGCTCCTCTTC[A/T]GACTTATCATTCTTCATCTCTACTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 164020 | ||||||||||||||||||||
Literature Links: |
HNRNPC PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HNRNPC - heterogeneous nuclear ribonucleoprotein C (C1/C2) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001077442.1 | 904 | Silent Mutation | TCA,TCT | S,S 233 | NP_001070910.1 | |
NM_001077443.1 | 904 | Silent Mutation | TCA,TCT | S,S 220 | NP_001070911.1 | |
NM_004500.3 | 904 | Silent Mutation | TCA,TCT | S,S 220 | NP_004491.2 | |
NM_031314.2 | 904 | Silent Mutation | TCA,TCT | S,S 233 | NP_112604.2 | |
XM_006720125.2 | 904 | Silent Mutation | TCA,TCT | S,S 220 | XP_006720188.1 | |
XM_011536708.1 | 904 | Silent Mutation | TCA,TCT | S,S 233 | XP_011535010.1 | |
XM_011536709.2 | 904 | Silent Mutation | TCA,TCT | S,S 233 | XP_011535011.1 | |
XM_011536710.2 | 904 | Silent Mutation | TCA,TCT | S,S 233 | XP_011535012.1 | |
XM_011536711.2 | 904 | Silent Mutation | TCA,TCT | S,S 233 | XP_011535013.1 | |
XM_011536712.1 | 904 | Silent Mutation | TCA,TCT | S,S 220 | XP_011535014.1 | |
XM_017021252.1 | 904 | Silent Mutation | TCA,TCT | S,S 220 | XP_016876741.1 | |
XM_017021253.1 | 904 | Silent Mutation | TCA,TCT | S,S 220 | XP_016876742.1 |
LINC00641 - long intergenic non-protein coding RNA 641 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |