Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTTTCCATTTATTTGCAGGTGTGG[A/T]TTTGCTGGAGAAACTGGTCCAAGAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ACTR10 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ACTR10 - actin-related protein 10 homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018477.2 | 143 | Silent Mutation | GGA,GGT | G,G 28 | NP_060947.1 | |
XM_011536960.1 | 143 | Silent Mutation | GGA,GGT | G,G 28 | XP_011535262.1 | |
XM_011536961.1 | 143 | Silent Mutation | GGA,GGT | G,G 28 | XP_011535263.1 | |
XM_011536962.2 | 143 | UTR 5 | XP_011535264.1 | |||
XM_011536963.2 | 143 | UTR 5 | XP_011535265.1 |