Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATAGGAGACGATGCCGTGGGCCACA[C/T]TGTTACACAGCAGGGGGCCTCCGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 116830 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CTSG PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CTSG - cathepsin G | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001911.2 | 663 | Missense Mutation | AAT,AGT | N,S 209 | NP_001902.1 | |
XM_011536499.1 | 663 | Missense Mutation | AAT,AGT | N,S 223 | XP_011534801.1 |