Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGTTCGCCTCCGGAGCCATGGCGG[C/T]GGCACTGAAGTGTCTACTGACATTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616091 MIM: 612166 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC101929718 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC101929718 - uncharacterized LOC101929718 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
METTL17 - methyltransferase like 17 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001029991.1 | 205 | Missense Mutation | GCG,GTG | A,V 3 | NP_001025162.1 | |
NM_022734.2 | 205 | Missense Mutation | GCG,GTG | A,V 3 | NP_073571.1 | |
XM_006720236.2 | 205 | Intron | XP_006720299.1 | |||
XM_017021601.1 | 205 | Intron | XP_016877090.1 |
SLC39A2 - solute carrier family 39 member 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |