Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCCATCTTTCTCTTTGTCTTCCTC[A/C]TCATCATCATCATAGTCTTCTTCTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600520 | ||||||||||||||||||||
Literature Links: |
GTF2A1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GTF2A1 - general transcription factor IIA subunit 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278940.1 | 1386 | Missense Mutation | GAG,GAT | E,D 245 | NP_001265869.1 | |
NM_015859.3 | 1386 | Missense Mutation | GAG,GAT | E,D 295 | NP_056943.1 | |
NM_201595.2 | 1386 | Missense Mutation | GAG,GAT | E,D 256 | NP_963889.1 |