Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACCCCGGGCAGGCACCCCCAGGGG[A/C]TTATCCTGGACAGGCACCTCCAGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 153619 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DLGAP5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DLGAP5 - DLG associated protein 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LGALS3 - lectin, galactoside binding soluble 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001177388.1 | 246 | Missense Mutation | GAT,GCT | D,A 53 | NP_001170859.1 | |
NM_002306.3 | 246 | Missense Mutation | GAT,GCT | D,A 53 | NP_002297.2 | |
XM_011536759.2 | 246 | Missense Mutation | GAT,GCT | D,A 67 | XP_011535061.2 |