Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCTTACCGCCCAGTACGCAGACTC[C/T]GTGAGTACCCGGGAGAGATCAGGGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603372 | ||||||||||||||||||||
Literature Links: |
CEP128 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CEP128 - centrosomal protein 128 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152446.3 | 546 | Intron | NP_689659.2 | |||
XM_006720056.3 | 546 | Intron | XP_006720119.1 | |||
XM_011536490.2 | 546 | Intron | XP_011534792.1 | |||
XM_011536491.2 | 546 | Intron | XP_011534793.1 | |||
XM_011536492.2 | 546 | Intron | XP_011534794.1 | |||
XM_011536493.2 | 546 | Intron | XP_011534795.1 | |||
XM_011536495.2 | 546 | Intron | XP_011534797.1 | |||
XM_017021042.1 | 546 | Intron | XP_016876531.1 | |||
XM_017021043.1 | 546 | Intron | XP_016876532.1 |
TSHR - thyroid stimulating hormone receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000369.2 | 546 | Missense Mutation | CCG,CTG | P,L 57 | NP_000360.2 | |
NM_001018036.2 | 546 | Missense Mutation | CCG,CTG | P,L 57 | NP_001018046.1 | |
NM_001142626.2 | 546 | Missense Mutation | CCG,CTG | P,L 57 | NP_001136098.1 | |
XM_005268037.4 | 546 | Missense Mutation | CCG,CTG | P,L 57 | XP_005268094.1 | |
XM_005268039.1 | 546 | Missense Mutation | CCG,CTG | P,L 57 | XP_005268096.1 | |
XM_006720245.1 | 546 | Missense Mutation | CCG,CTG | P,L 57 | XP_006720308.1 | |
XM_011537119.2 | 546 | UTR 5 | XP_011535421.1 |