Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCGGGAGGCGTCGTCCTCCAGCCCC[C/G]AGGCCGGCGAAGGGCAGATCCGCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615317 MIM: 602091 MIM: 601015 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ISCA2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ISCA2 - iron-sulfur cluster assembly 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001272007.1 | 185 | Missense Mutation | CAG,GAG | Q,E 44 | NP_001258936.1 | |
NM_194279.3 | 185 | Missense Mutation | CAG,GAG | Q,E 44 | NP_919255.2 |
LTBP2 - latent transforming growth factor beta binding protein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NPC2 - NPC intracellular cholesterol transporter 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006432.3 | 185 | Intron | NP_006423.1 | |||
XM_017020930.1 | 185 | Intron | XP_016876419.1 |