Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGATGAAAAAGGAGAAAAGCCACCA[C/T]ATCCATCTCATGAAAGATGAGTGCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603930 MIM: 607849 | ||||||||||||||||||||
Literature Links: |
GPHN PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RDH11 - retinol dehydrogenase 11 (all-trans/9-cis/11-cis) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001252650.1 | 837 | Missense Mutation | ATA,ATG | I,M 179 | NP_001239579.1 | |
NM_016026.3 | 837 | Missense Mutation | ATA,ATG | I,M 249 | NP_057110.3 |