Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATGTGGAAGTAGAGAGTCTTGTTG[C/T]GCAGCAGGGAGTAGGAGTTGTCGAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
TMED8 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TMED8 - transmembrane p24 trafficking protein family member 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_213601.1 | 837 | Missense Mutation | CAC,CGC | H,R 313 | NP_998766.1 | |
XM_017021223.1 | 837 | Missense Mutation | CAC,CGC | H,R 420 | XP_016876712.1 | |
XM_017021224.1 | 837 | Missense Mutation | CAC,CGC | H,R 250 | XP_016876713.1 | |
XM_017021225.1 | 837 | Missense Mutation | CAC,CGC | H,R 245 | XP_016876714.1 |