Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCAGAGTTCCCAGGCAGAGCCATC[A/G]GCCCAGGCCAGGACAGTAGGCATGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
KLHL33 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KLHL33 - kelch like family member 33 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001109997.2 | 2699 | Nonsense Mutation | CGA,TGA | R,* 493 | NP_001103467.2 | |
XM_005267347.4 | 2699 | Nonsense Mutation | CGA,TGA | R,* 757 | XP_005267404.1 | |
XM_011536450.2 | 2699 | Nonsense Mutation | CGA,TGA | R,* 757 | XP_011534752.1 |