Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTAACCCCGAAACAGCTACTGGG[G/T]ATAGCTGGAAGATTTGAATATCTCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615094 | ||||||||||||||||||||
Literature Links: |
PROX2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PROX2 - prospero homeobox 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080408.2 | 2849 | Missense Mutation | ACC,CCC | T,P 364 | NP_001073877.2 | |
NM_001243007.1 | 2849 | Missense Mutation | ACC,CCC | T,P 591 | NP_001229936.1 | |
XM_005267543.3 | 2849 | Missense Mutation | ACC,CCC | T,P 591 | XP_005267600.1 | |
XM_017021222.1 | 2849 | Intron | XP_016876711.1 |