Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCTCCGGAAGAGGGAGCCCAGTCC[T/C]CGCCACCGCCGAGTCTGCGGAACCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610861 | ||||||||||||||||||||
Literature Links: |
LINC00341 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LINC00341 - long intergenic non-protein coding RNA 341 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SYNE3 - spectrin repeat containing nuclear envelope family member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152592.3 | 2741 | Silent Mutation | CGA,CGG | R,R 914 | NP_689805.3 | |
XM_005267377.3 | 2741 | Silent Mutation | CGA,CGG | R,R 914 | XP_005267434.1 | |
XM_006720063.3 | 2741 | Silent Mutation | CGA,CGG | R,R 914 | XP_006720126.1 | |
XM_011536515.2 | 2741 | Silent Mutation | CGA,CGG | R,R 922 | XP_011534817.1 | |
XM_017021056.1 | 2741 | Silent Mutation | CGA,CGG | R,R 909 | XP_016876545.1 | |
XM_017021057.1 | 2741 | Intron | XP_016876546.1 |