Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATGGTCGGGACTGAGGTGCTAGAG[C/G]GGGGAGTGAATTGGCCACTTGTTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600037 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
OTX2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
OTX2 - orthodenticle homeobox 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270523.1 | 838 | Missense Mutation | CCC,CGC | P,R 134 | NP_001257452.1 | |
NM_001270524.1 | 838 | Missense Mutation | CCC,CGC | P,R 134 | NP_001257453.1 | |
NM_001270525.1 | 838 | Missense Mutation | CCC,CGC | P,R 142 | NP_001257454.1 | |
NM_021728.3 | 838 | Missense Mutation | CCC,CGC | P,R 142 | NP_068374.1 | |
NM_172337.2 | 838 | Missense Mutation | CCC,CGC | P,R 134 | NP_758840.1 |