Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTATGGATTCTGGACACTTCTTGG[C/T]GGGAATGGTCCGTGGCCAGCACGTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606167 | ||||||||||||||||||||
Literature Links: |
GPR132 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GPR132 - G protein-coupled receptor 132 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278694.1 | 1085 | Missense Mutation | CAC,CGC | H,R 316 | NP_001265623.1 | |
NM_001278695.1 | 1085 | Missense Mutation | CAC,CGC | H,R 307 | NP_001265624.1 | |
NM_001278696.1 | 1085 | Missense Mutation | CAC,CGC | H,R 128 | NP_001265625.1 | |
NM_013345.3 | 1085 | Missense Mutation | CAC,CGC | H,R 316 | NP_037477.1 |