Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCCTCTGGGTAGCAGCTGTGCCCC[A/G]TGAGCGGGGCTCTCTCGGACAGGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602620 | ||||||||||||||||||||
Literature Links: |
LGMN PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LGMN - legumain | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001008530.2 | 1279 | Missense Mutation | ACG,ATG | T,M 374 | NP_001008530.1 | |
NM_005606.6 | 1279 | Missense Mutation | ACG,ATG | T,M 374 | NP_005597.3 | |
XM_005267862.3 | 1279 | Intron | XP_005267919.1 | |||
XM_005267863.3 | 1279 | Intron | XP_005267920.1 | |||
XM_011536970.1 | 1279 | Missense Mutation | ACG,ATG | T,M 374 | XP_011535272.1 | |
XM_017021463.1 | 1279 | Intron | XP_016876952.1 | |||
XM_017021464.1 | 1279 | Missense Mutation | ACG,ATG | T,M 374 | XP_016876953.1 | |
XM_017021465.1 | 1279 | Intron | XP_016876954.1 | |||
XM_017021466.1 | 1279 | Intron | XP_016876955.1 | |||
XM_017021467.1 | 1279 | Intron | XP_016876956.1 |