Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGTTGAAAACGCCCTTGGCGAAGT[C/T]GCAGCTGGCCTCCCAGACGCTGCGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601404 | ||||||||||||||||||||
Literature Links: |
C14orf159 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C14orf159 - chromosome 14 open reading frame 159 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GPR68 - G protein-coupled receptor 68 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001177676.1 | 1071 | Missense Mutation | AAC,GAC | N,D 259 | NP_001171147.1 | |
NM_003485.3 | 1071 | Missense Mutation | AAC,GAC | N,D 259 | NP_003476.3 | |
XM_005268110.4 | 1071 | Missense Mutation | AAC,GAC | N,D 269 | XP_005268167.1 | |
XM_005268111.3 | 1071 | Missense Mutation | AAC,GAC | N,D 269 | XP_005268168.1 | |
XM_005268112.3 | 1071 | Missense Mutation | AAC,GAC | N,D 269 | XP_005268169.1 | |
XM_006720262.3 | 1071 | Missense Mutation | AAC,GAC | N,D 269 | XP_006720325.1 | |
XM_011537196.2 | 1071 | Missense Mutation | AAC,GAC | N,D 269 | XP_011535498.1 | |
XM_011537197.2 | 1071 | Missense Mutation | AAC,GAC | N,D 269 | XP_011535499.1 | |
XM_011537198.2 | 1071 | Missense Mutation | AAC,GAC | N,D 269 | XP_011535500.1 | |
XM_011537199.2 | 1071 | Missense Mutation | AAC,GAC | N,D 269 | XP_011535501.1 |