Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACCCGCCCATCTGCGTGTCCGACG[A/G]CAACCGCCTGTGCACCCTGGCGTCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601038 MIM: 608523 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DIO3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DIO3 - deiodinase, iodothyronine, type III | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001362.3 | 460 | Missense Mutation | GAC,GGC | D,G 105 | NP_001353.4 |
DIO3OS - DIO3 opposite strand/antisense RNA (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR1247 - microRNA 1247 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |